Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-4-methylnicotinic acid ≥95%

Supplier: AOB CHEM USA
Warning

92283-500MG 92283-1G 92283-5G
77338-280EA 470.71 USD InStock
77338-280 77359-236 77383-204
5-Chloro-4-methylnicotinic acid ≥95%
5-Chloro-4-methylnicotinic acid
Precaución: for Research Use Only


Formula: C₇H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD07439972
CAS Number: 890092-41-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty