Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloro-6-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOB CHEM USA
Warning

98959-500MG 98959-1G 98959-5G
77336-980EA 401.7 USD InStock
77336-980 77359-340 77383-328
3-Bromo-2-chloro-6-(trifluoromethoxy)benzoic acid ≥97%
3-Bromo-2-chloro-6-(trifluoromethoxy)benzoic acid
Precaución: for Research Use Only


Formula: C₈H₃BrClF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD34775849
CAS Number: 2167664-93-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty