Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chlorobenzyl Alcohol ≥97%

Supplier: AOB CHEM USA
Warning

65752-5G 65752-25G
77368-664EA 142.14 USD InStock
77368-664 77380-868
3-Bromo-2-chlorobenzyl Alcohol ≥97%
3-Bromo-2-chlorobenzyl Alcohol


Formula: C₇H₆BrClO
Storage Temperature: Ambient
MDL Number: MFCD12923279
CAS Number: 1261524-75-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty