Olsen-Hegar Needle Holders, Sklar
Supplier: Sklar
20-2748 20-2755 20-2765 20-2775 20-2747
82030-126EA 198.64 USD InStock
82030-126 82030-128 82030-130 82030-132 82030-124
Olsen-Hegar Needle Holders, Sklar
Forceps
Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built in scissors
It is used with intermediate weight sutures, commonly in plastic surgery procedures.
Información para pedidos: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Precaución: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty