Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetra(ethylene glycol) dithiol 98%

Supplier: AMBEED, INC
Danger

A1465563-1G A1465563-250MG
77441-312EA 219 USD InStock
77441-312 77441-314 77441-316
Tetra(ethylene glycol) dithiol 98%
Tetra(ethylene glycol) dithiol
Precaución: for Research Use Only


Formula: C₈H₁₈O₃S₂
MDL Number: MFCD18910705
CAS Number: 2781-02-4
UN: 3334
ADR: 9,
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty