Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)benzyl alcohol 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR0847-1G OR0847-250MG OR0847-5G
77402-264EA 90.68 USD InStock
77402-264 77402-266 77402-268
4-(Dimethylamino)benzyl alcohol 97%
4-(Dimethylamino)benzyl alcohol


Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 125…126 °C
Storage Temperature: Freezer
MDL Number: MFCD03840449
CAS Number: 1703-46-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty