Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Periodic acid 99+%

Supplier: APOLLO SCIENTIFIC
Danger

IN2845-50G IN2845-5G
77401-526EA 124.3 USD InStock
77401-526 77401-528
Periodic acid 99+%
Periodic acid


Formula: H₅IO₆
MW: 227.94 g/mol
Boiling Pt: 140 °C
Melting Pt: 122
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011344
CAS Number: 10450-60-9
EINECS: 233-937-0
UN: 3085
ADR: 5.1+8 II
Merck Index: 13,07248
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty