Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Naphth-1-yl)piperazine dihydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR0079-1G OR0079-250MG
77401-676EA 195.62 USD InStock
77401-676 77401-678
1-(Naphth-1-yl)piperazine dihydrochloride 97%
1-(Naphth-1-yl)piperazine dihydrochloride


Formula: C₁₄H₁₆N₂·2ClH
Melting Pt: >260 °C
Storage Temperature: Ambient
MDL Number: MFCD03540959
CAS Number: 1188264-04-1
UN: 1759
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty