Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzo[b]thiophene-2-boronic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR0093-1G OR0093-25G OR0093-5G
77401-686EA 21.4 USD InStock
77401-686 77401-688 77401-690
Benzo[b]thiophene-2-boronic acid
Benzo[b]thiophene-2-boronic acid


Formula: C₈H₇BO₂S
MW: 178.02
Melting Pt: 252…255 °C
Storage Temperature: Freezer
MDL Number: MFCD01075674
CAS Number: 98437-23-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty