Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR015663-1G OR015663-500MG
77401-764EA 584.8 USD InStock
77401-764 77401-766
3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide 95%
3-Chloro-4-hydroxytetrahydrothiophene 1,1-dioxide


Formula: C₄H₇ClO₃S
MW: 170.615 g/mol
Melting Pt: 163…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005486
CAS Number: 49592-61-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty