Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-2,4-dimethylacetophenone 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR015117-1G OR015117-25G OR015117-5G
77401-744EA 34.64 USD InStock
77401-744 77401-746 77401-748
ω-Bromo-2,4-dimethylacetophenone 97%
ω-Bromo-2,4-dimethylacetophenone


Formula: C₁₀H₁₁BrO
MW: 227.1
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00017872
CAS Number: 26346-85-0
UN: 3261
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty