Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(4-isobutylphenyl)propanoate

Supplier: APOLLO SCIENTIFIC
Warning

OR0631-1G OR0631-250MG
77402-164EA 28.36 USD InStock
77402-164 77402-166
Methyl 2-(4-isobutylphenyl)propanoate
Methyl 2-(4-isobutylphenyl)propanoate


Formula: C₁₄H₂₀O₂
Boiling Pt: 100 °C (0.8 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00869922
CAS Number: 61566-34-5
EINECS: 262-853-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty