Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Neodymium(III) acetate hydrate 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

IN2623-250G IN2623-50G
77401-492EA 412.62 USD InStock
77401-492 77401-494
Neodymium(III) acetate hydrate 99,9%
Neodymium(III) acetate hydrate


Formula: C₆H₁₁NdO₇
MW: 339.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013040
CAS Number: 334869-71-5
EINECS: 228-244-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty