Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gallium tribromide, anhydrous 99.99%

Supplier: APOLLO SCIENTIFIC

IN1867-100G IN1867-25G
77401-454EA 1450.8 USD InStock
77401-454 77401-456
Gallium tribromide, anhydrous 99.99%
Gallium tribromide


Formula: Br₃Ga
Boiling Pt: 279 °C
Melting Pt: 121.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00016100
CAS Number: 13450-88-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty