Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Iron(III) bromide, anhydrous

Supplier: APOLLO SCIENTIFIC
Warning

IN2056-100G IN2056-25G IN2056-5G
77401-466EA 132.44 USD InStock
77401-466 77401-468 77401-470
Iron(III) bromide, anhydrous
Iron(III) bromide


Formula: Br₃Fe
MW: 295.56 g/mol
Melting Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00016082
CAS Number: 10031-26-2
EINECS: 233-089-1
Merck Index: 12,04060
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty