Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium hexacyanoruthenate(II) trihydrate 99%

Supplier: APOLLO SCIENTIFIC
Warning

IN2911-1G IN2911-5G
77401-534EA 206.82 USD InStock
77401-534 77401-536
Potassium hexacyanoruthenate(II) trihydrate 99%
Potassium hexacyanoruthenate(II) trihydrate


Formula: C₆K₄N₆Ru·3H₂O
MW: 467.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00168064
CAS Number: 15002-31-0
UN: 1588
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty