Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Barium thiocyanate trihydrate 99%

Supplier: APOLLO SCIENTIFIC

IN1225-100G IN1225-25G
77401-366EA 241.46 USD InStock
77401-366 77401-368
Barium thiocyanate trihydrate 99%
Barium thiocyanate trihydrate


Formula: Ba(SCN)₂·3H₂O
MW: 307.55 g/mol (253.49 anhy)
Density: 2.286 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150182
CAS Number: 68016-36-4
UN: 1564
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty