Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Indole-5-carbaldehyd 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10488-25G OR10488-5G
77402-738EA 187.46 USD InStock
77402-738 77402-740
Indole-5-carbaldehyd 98%
Indole-5-carbaldehyd


Formula: C₉H₇NO
MW: 145.16 g/mol
Melting Pt: 96…103 °C
Storage Temperature: Ambient
MDL Number: MFCD02093664
CAS Number: 1196-69-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty