Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diazaspiro[4.5]decane-2,4-dione

Supplier: APOLLO SCIENTIFIC
Warning

OR0331-250MG OR0331-5G OR0331-1G
77402-034EA 21.4 USD InStock
77402-034 77402-036 77402-032
1,3-Diazaspiro[4.5]decane-2,4-dione
1,3-Diazaspiro[4.5]decane-2,4-dione


Formula: C₈H₁₂N₂O₂
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00022404
CAS Number: 702-62-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty