Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-6-methoxypyrimidin-2-amine 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR10959-100G OR10959-25G
77402-996EA 28.28 USD InStock
77402-996 77402-998
4-Chloro-6-methoxypyrimidin-2-amine 96%
4-Chloro-6-methoxypyrimidin-2-amine


Formula: C₅H₆ClN₃O
Melting Pt: 168…176 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00233535
CAS Number: 5734-64-5
EINECS: 410-050-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty