Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-nitroaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR015002-100G OR015002-25G OR015002-5G
77401-716EA 103.92 USD InStock
77401-716 77401-718 77401-720
2-Chloro-4-nitroaniline 98%
2-Chloro-4-nitroaniline


Formula: C₆H₅ClN₂O₂
Melting Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00007665
CAS Number: 121-87-9
UN: 2237
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty