Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Cyclohexylmethyl)hydrazine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR0199-10G OR0199-1G OR0199-5G
77401-922EA 660.2 USD InStock
77401-922 77401-924 77401-936
(Cyclohexylmethyl)hydrazine hydrochloride 97%
(Cyclohexylmethyl)hydrazine hydrochloride


Formula: C₇H₁₆N₂·ClH
Melting Pt: 141…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06245509
CAS Number: 3637-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty