Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dioxaspiro[4.5]decan-8-one

Supplier: APOLLO SCIENTIFIC
Warning

OR0427-100G OR0427-500G
77402-084EA 66.22 USD InStock
77402-084 77402-086
1,4-Dioxaspiro[4.5]decan-8-one
1,4-Dioxaspiro[4.5]decan-8-one


Formula: C₈H₁₂O₃
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00010214
CAS Number: 4746-97-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty