Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L(+)-Diethyl tartrate

Supplier: APOLLO SCIENTIFIC
Warning

OR10049-100G OR10049-500G
77402-416EA 29.54 USD InStock
77402-416 77402-418
L(+)-Diethyl tartrate
L(+)-Diethyl tartrate
Asymmetric synthesis catalyst; Chiral intermediate for pharmaceuticals and agrochemicals.


Formula: C₈H₁₄O₆
MW: 206.2 g/mol
Boiling Pt: 280
Melting Pt: 18.7
Density: 1.204 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00009143
CAS Number: 87-91-2
EINECS: 201-783-3
Merck Index: 13,03886
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty