Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR11129-10G OR11129-5G
77403-712EA 59.1 USD InStock
77403-712 77403-716
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane 96%
1,1-Dibromo-2,2-bis(chloromethyl)cyclopropane


Formula: C₅H₆Br₂Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00101445
CAS Number: 98577-44-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty