Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Piperidin-4-ylmethylpyridine dihydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR12878-100MG OR12878-250MG
77404-458EA 350.48 USD InStock
77404-458 77404-460
2-Piperidin-4-ylmethylpyridine dihydrochloride
2-Piperidin-4-ylmethylpyridine dihydrochloride


Formula: C₁₁H₁₈Cl₂N₂
Storage Temperature: Ambient
MDL Number: MFCD03840134
CAS Number: 886886-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty