Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Methyl-4-piperidone 98+%

Supplier: APOLLO SCIENTIFIC
Danger

OR0648-100G OR0648-25G OR0648-5G
77402-174EA 89.66 USD InStock
77402-174 77402-176 77402-178
1-Methyl-4-piperidone 98+%
1-Methyl-4-piperidone


Formula: C₆H₁₁NO
Boiling Pt: 55…58 °C/10 mmHg
Density: 0.973 g/cm³ (25 °C)
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5
UN: 2733
ADR: 3+8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty