Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxysalicylaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR0913-100G OR0913-25G OR0913-500G
77402-302EA 114.1 USD InStock
77402-302 77402-304 77402-306
4-Methoxysalicylaldehyde 97%
4-Methoxysalicylaldehyde


Formula: C₈H₈O₃
Melting Pt: 40…43 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00003327
CAS Number: 673-22-3
EINECS: 211-604-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty