Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3,3-Dimethoxy-1-methylpropylidene)malononitrile

Supplier: APOLLO SCIENTIFIC
Danger

OR110842-1G OR110842-250MG
77403-500EA 835.06 USD InStock
77403-500 77403-502
(3,3-Dimethoxy-1-methylpropylidene)malononitrile
(3,3-Dimethoxy-1-methylpropylidene)malononitrile


Formula: C₉H₁₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09030793
CAS Number: 410547-37-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty