Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methoxythiophene-2-carbonitrile

Supplier: APOLLO SCIENTIFIC
Danger

OR110878-250MG OR110878-5G OR110878-1G
77403-534EA 54.8 USD InStock
77403-534 77403-536 77403-532
3-Methoxythiophene-2-carbonitrile
3-Methoxythiophene-2-carbonitrile


Formula: C₆H₅NOS
MW: 139.175 g/mol
Boiling Pt: 72…74 °C (0.1 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD24052834
CAS Number: 57059-18-4
UN: 2810
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty