Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-Heptenylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR10493-1G OR10493-5G
77402-746EA 158.94 USD InStock
77402-746 77402-748
trans-Heptenylboronic acid 97%
trans-Heptenylboronic acid


Formula: C₇H₁₅BO₂
MW: 142.01 g/mol
Melting Pt: 107…111 °C
Storage Temperature: Freezer
MDL Number: MFCD01074600
CAS Number: 57404-76-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty