Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR110758-1G OR110758-250MG
77403-478EA 157.94 USD InStock
77403-478 77403-480
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid 95%
3-Oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylic acid


Formula: C₉H₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD07774185
CAS Number: 214848-62-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty