Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chloroquinazoline

Supplier: APOLLO SCIENTIFIC
Warning

OR110444-10G OR110444-1G OR110444-5G
77403-338EA 267.94 USD InStock
77403-338 77403-340 77403-342
2-Amino-5-chloroquinazoline
2-Amino-5-chloroquinazoline


Formula: C₈H₆ClN₃
Melting Pt: 249…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00974335
CAS Number: 190273-70-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty