Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethyl-1-butyne

Supplier: APOLLO SCIENTIFIC
Danger

OR11141-100G OR11141-25G OR11141-500G
77403-758EA 447.26 USD InStock
77403-758 77403-760 77403-762
3,3-Dimethyl-1-butyne
3,3-Dimethyl-1-butyne


Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 38
Melting Pt: -78.2
Density: 0.671 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008852
CAS Number: 917-92-0
EINECS: 213-035-3
UN: 3295
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty