Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6-bromopicolinate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11544-25G OR11544-5G
77403-942EA 103.92 USD InStock
77403-942 77403-944
Ethyl-6-bromopicolinate 98%
Ethyl 6-bromopicolinate


Formula: C₈H₈BrNO₂
Melting Pt: 41 °C
Storage Temperature: Ambient
MDL Number: MFCD04116913
CAS Number: 21190-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty