Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxypyridine

Supplier: APOLLO SCIENTIFIC
Warning

OR1212-100G OR1212-25G
77404-192EA 154.86 USD InStock
77404-192 77404-194
5-Bromo-2-methoxypyridine
5-Bromo-2-methoxypyridine


Formula: C₆H₆BrNO
Boiling Pt: 80 °C/12 mmHg
Density: 1.453 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01318952
CAS Number: 13472-85-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty