Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(2,3-Dichlorophenyl)-4-oxazolecarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR11671-100MG OR11671-500MG
77404-004EA 63.16 USD InStock
77404-004 77404-006
5-(2,3-Dichlorophenyl)-4-oxazolecarboxylic acid
5-(2,3-Dichlorophenyl)-4-oxazolecarboxylic acid


Formula: C₁₀H₅Cl₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD07376179
CAS Number: 951885-34-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty