Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-(3-hydroxyphenyl)propanoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR13244-10G OR13244-1G OR13244-5G
77404-604EA 330.1 USD InStock
77404-604 77404-606 77404-608
3-Amino-3-(3-hydroxyphenyl)propanoic acid
3-Amino-3-(3-hydroxyphenyl)propanoic acid


Formula: C₉H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD01871309
CAS Number: 102872-33-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty