Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3',5'-Dimethyl-4'-hydroxyacetophenone

Supplier: APOLLO SCIENTIFIC
Warning

OR13892-1G OR13892-5G OR13892-10G
77404-754EA 117.16 USD InStock
77404-754 77404-758 77404-752
3',5'-Dimethyl-4'-hydroxyacetophenone
3',5'-Dimethyl-4'-hydroxyacetophenone


Formula: C₁₀H₁₂O₂
Melting Pt: 151…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00195574
CAS Number: 5325-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty