Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenoxypropionic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR13898-1G OR13898-25G OR13898-5G
77404-768EA 55.02 USD InStock
77404-768 77404-770 77404-772
3-Phenoxypropionic acid
3-Phenoxypropionic acid


Formula: C₉H₁₀O₃
MW: 166.18
Boiling Pt: 235
Melting Pt: 92
Storage Temperature: Ambient
MDL Number: MFCD00002769
CAS Number: 7170-38-9
EINECS: 230-518-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty