Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiazole-2-carbaldehyde 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR14343-1G OR14343-250MG OR14343-5G
77404-914EA 23.28 USD InStock
77404-914 77404-916 77404-918
Thiazole-2-carbaldehyde 95+%
Thiazole-2-carbaldehyde


Formula: C₄H₃NOS
MW: 113.13 g/mol
Boiling Pt: 61…63 °C/15 mmHg
Density: 1.288 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142924
CAS Number: 10200-59-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty