Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Azetidin-3-ol hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR1873-100G OR1873-25G OR1873-500G
77406-372EA 61.12 USD InStock
77406-372 77406-374 77406-376
Azetidin-3-ol hydrochloride 97%
Azetidin-3-ol hydrochloride


Formula: C₃H₈ClNO
Melting Pt: 85…90 °C
Storage Temperature: Ambient
MDL Number: MFCD02683887
CAS Number: 18621-18-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty