Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridine-6-carbaldehyde 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR111513-1G OR111513-250MG OR111513-5G
77403-798EA 268.96 USD InStock
77403-798 77403-800 77403-802
Imidazo[1,2-a]pyridine-6-carbaldehyde 96%
Imidazo[1,2-a]pyridine-6-carbaldehyde


Formula: C₈H₆N₂O
MW: 146.15 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD07772808
CAS Number: 116355-16-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty