Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,4-Dibromotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR13169-100G OR13169-25G OR13169-500G
77404-566EA 103.92 USD InStock
77404-566 77404-568 77404-570
α,4-Dibromotoluene 98%
α,4-Dibromotoluene


Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 115…124 °C/12 mmHg
Melting Pt: 60
Storage Temperature: Ambient
MDL Number: MFCD00000179
CAS Number: 589-15-1
EINECS: 209-636-5
UN: 3261
ADR: 8 II
Merck Index: 13,01393
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty