Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Quinolinol 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR14331-10G OR14331-25G OR14331-5G
77404-908EA 83.54 USD InStock
77404-908 77404-910 77404-912
5-Quinolinol 98%
5-Quinolinol


Formula: C₉H₇NO
Melting Pt: 223…226 °C
Storage Temperature: Ambient
MDL Number: MFCD00006792
CAS Number: 578-67-6
EINECS: 209-428-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty