Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Hexen-1-ol 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR14506-1G OR14506-25G OR14506-5G
77404-986EA 21.26 USD InStock
77404-986 77404-988 77404-990
5-Hexen-1-ol 97%
5-Hexen-1-ol


Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 152…155 °C
Melting Pt: < –20 °C
Density: 0.834 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002981
CAS Number: 821-41-0
EINECS: 212-477-4
UN: 2924
ADR: 3+8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty