Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methylfuran-3-boronic acid pinacol ester 90%

Supplier: APOLLO SCIENTIFIC
Warning

OR14882-1G OR14882-250MG OR14882-5G
77405-134EA 190.52 USD InStock
77405-134 77405-136 77405-138
2-Methylfuran-3-boronic acid pinacol ester 90%
2-Methylfuran-3-boronic acid pinacol ester


Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD09879905
CAS Number: 864776-02-3
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty