Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ω-Bromo-2,4-dichloroacetophenone 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR12270-100G OR12270-25G OR12270-5G
77404-234EA 520.62 USD InStock
77404-234 77404-236 77404-238
ω-Bromo-2,4-dichloroacetophenone 95%
ω-Bromo-2,4-dichloroacetophenone


Formula: C₈H₅BrCl₂O
Melting Pt: 25…29 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00053005
CAS Number: 2631-72-3
EINECS: 220-116-7
UN: 1759
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty