Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-4-chloropicolinate 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR1352-100G OR1352-25G OR1352-5G
77404-656EA 64.18 USD InStock
77404-656 77404-658 77404-660
Methyl-4-chloropicolinate 97%
Methyl 4-chloropicolinate


Formula: C₇H₆ClNO₂
MW: 171.5800 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD04116183
CAS Number: 24484-93-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty