Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Isothiocyanatopyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR1254-1G OR1254-25G OR1254-5G
77404-344EA 61.12 USD InStock
77404-344 77404-346 77404-348
3-Isothiocyanatopyridine 98%
3-Isothiocyanatopyridine


Formula: C₆H₄N₂S
Boiling Pt: 231…233 °C
Density: 1.22 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00052351
CAS Number: 17452-27-6
UN: 2927
ADR: 6.1+8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty